Learn More
AdipoGen Ansatrienin A
SDP
Catalog No. 502562048
Encompass_Preferred
Chemical. CAS 82189-03-5. Formula C36H48N2O8. MW 636.8. Isolated from Streptomyces collinus. Antibiotic. Antitumor compound. Antifungal. Inhibits osteoclastic bone resorption.
Catalog No. 50-256-2048 Supplier AdipoGen Supplier No. BVT0246M001
Chemical. CAS 82189-03-5. Formula C36H48N2O8. MW 636.8. Isolated from Streptomyces collinus. Antibiotic. Antitumor compound. Antifungal. Inhibits osteoclastic bone resorption.