Learn More
MSC Ansell 97-003 Gloves, Cut A4, Size 2X-Large, 1 Pair
Catalog No. 5000686488
Encompass
The Ansell 97-003 gloves are designed for demanding applications requiring superior hand protection. They offer robust resistance against cuts, punctures, and abrasions, making them ideal for various industrial tasks.
  • Constructed from nylon, spandex, stainless steel and Kevlar
  • Features a foam nitrile-coated palm
  • Provides ANSI/ISEA Cut Resistance Level A4
  • Offers ANSI/ISEA Puncture Resistance Level 3
  • Delivers ANSI/ISEA Abrasion Resistance Level 4
  • Features 13 gauge for optimal dexterity and protection
  • Equipped with a secure Velcro cuff
  • EN 388 rated 4X31D
Catalog No. 50-006-86488 Supplier MSC Supplier No. 73779878
The Ansell 97-003 gloves are designed for demanding applications requiring superior hand protection. They offer robust resistance against cuts, punctures, and abrasions, making them ideal for various industrial tasks.
  • Constructed from nylon, spandex, stainless steel and Kevlar
  • Features a foam nitrile-coated palm
  • Provides ANSI/ISEA Cut Resistance Level A4
  • Offers ANSI/ISEA Puncture Resistance Level 3
  • Delivers ANSI/ISEA Abrasion Resistance Level 4
  • Features 13 gauge for optimal dexterity and protection
  • Equipped with a secure Velcro cuff
  • EN 388 rated 4X31D