Learn More
MSC Ansell ActivArmr Gloves, Cut A5, 13 ga, Small, 1 Pair
Catalog No. 5000767381
Encompass
The Ansell ActivArmr 80-505 gloves provide exceptional cut resistance for general purpose and mechanic's applications. Constructed with a 13 gauge blend of Kevlar, Steel, Basalt, Polyester & Spandex, these hand-specific gloves feature a Foam Nitrile-coated palm for a sandy grip and operate up to 284 °F.
  • ANSI/ISEA Cut Resistance Level A5
  • Heat-resistant properties
  • Overall length of 8-7/8 in.
  • Elastic knit cuff
  • Unisex design, Small size
  • Foam Nitrile coating on palm and fingertips
Catalog No. 50-007-67381 Supplier MSC Supplier No. 56355878
The Ansell ActivArmr 80-505 gloves provide exceptional cut resistance for general purpose and mechanic's applications. Constructed with a 13 gauge blend of Kevlar, Steel, Basalt, Polyester & Spandex, these hand-specific gloves feature a Foam Nitrile-coated palm for a sandy grip and operate up to 284 °F.
  • ANSI/ISEA Cut Resistance Level A5
  • Heat-resistant properties
  • Overall length of 8-7/8 in.
  • Elastic knit cuff
  • Unisex design, Small size
  • Foam Nitrile coating on palm and fingertips