Learn More
MSC Ansell Chemical Resistant Gloves, AlphaTEC 58-201, Large, Nitrile, 1 ea
Catalog No. 5000793219
Encompass
This product consists of green, supported chemical resistant gloves, model AlphaTEC™ 58-201. They feature a nitrile coating and an acrylic lined nitrile dipped construction, providing a thickness of 80 mil. These heavy-duty gloves are designed to resist a range of chemicals, conforming to EN ISO 374-1-2016: Type A JKLOPT standards, and are suitable for demanding applications.
  • Supported design for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Gauntlet cuff style
  • Acrylic lined for comfort and protection
  • Heavy-duty construction for demanding use
Catalog No. 50-007-93219 Supplier MSC Supplier No. 67605782
This product consists of green, supported chemical resistant gloves, model AlphaTEC™ 58-201. They feature a nitrile coating and an acrylic lined nitrile dipped construction, providing a thickness of 80 mil. These heavy-duty gloves are designed to resist a range of chemicals, conforming to EN ISO 374-1-2016: Type A JKLOPT standards, and are suitable for demanding applications.
  • Supported design for additional strength
  • Resistant to snags, abrasions, cuts, and punctures
  • Gauntlet cuff style
  • Acrylic lined for comfort and protection
  • Heavy-duty construction for demanding use