Learn More
MSC Ansell Chemical Resistant Gloves, Natural Rubber Latex, 18 mil, X-Large, Unsupported
Catalog No. 5000507125
Encompass
These chemical resistant gloves are designed for tasks requiring dexterity and light-to-medium chemical protection. The unsupported design offers an economical solution for various applications.
  • Beaded cuff for tear resistance and easy donning
  • Food approved
  • Good abrasion resistance
  • 100% cotton flock lining
  • Reversed lozenge grip surface
  • Seamless construction
  • Meets REACH, EN ISO 21420:2020, CE 0493, CAT III CE 2777, EN ISO 374-5:2016, and EN 388:2016 +A1:2018 standards
Catalog No. 50-005-07125 Supplier MSC Supplier No. 93659027
These chemical resistant gloves are designed for tasks requiring dexterity and light-to-medium chemical protection. The unsupported design offers an economical solution for various applications.
  • Beaded cuff for tear resistance and easy donning
  • Food approved
  • Good abrasion resistance
  • 100% cotton flock lining
  • Reversed lozenge grip surface
  • Seamless construction
  • Meets REACH, EN ISO 21420:2020, CE 0493, CAT III CE 2777, EN ISO 374-5:2016, and EN 388:2016 +A1:2018 standards