Learn More
MSC Ansell Chemical-Resistant Gloves, Neoprene, 80 mil, 31.5 in, 1 Pair
Catalog No. 5000677702
Encompass
AlphaTec® 09-430 are long-cuffed neoprene chemical-resistant gloves designed to offer advanced mechanical and chemical protection. They feature a high-grade neoprene compound and smooth finish to withstand various hazards, including acids, oils, caustics, alcohols, and solvents, and a cotton jersey liner for comfort and protection against snags and punctures.
  • High-grade neoprene construction provides advanced chemical resistance.
  • Cotton jersey liner offers comfort and protection against snags and punctures.
  • Anatomical design promotes greater dexterity and reduced hand fatigue.
  • Withstands various mechanical and chemical hazards, including acids, oils, caustics, alcohols, and solvents.
  • Measures 800 mm (31.5 in.) ensuring comprehensive personal protection up to the wearer's shoulder.
Catalog No. 50-006-77702 Supplier MSC Supplier No. 07912298
AlphaTec® 09-430 are long-cuffed neoprene chemical-resistant gloves designed to offer advanced mechanical and chemical protection. They feature a high-grade neoprene compound and smooth finish to withstand various hazards, including acids, oils, caustics, alcohols, and solvents, and a cotton jersey liner for comfort and protection against snags and punctures.
  • High-grade neoprene construction provides advanced chemical resistance.
  • Cotton jersey liner offers comfort and protection against snags and punctures.
  • Anatomical design promotes greater dexterity and reduced hand fatigue.
  • Withstands various mechanical and chemical hazards, including acids, oils, caustics, alcohols, and solvents.
  • Measures 800 mm (31.5 in.) ensuring comprehensive personal protection up to the wearer's shoulder.