Learn More
MSC Ansell Cut and Abrasion-Resistant Gloves, Cut A3, X-Small, 1 Pair
Catalog No. 5000794581
Encompass
These cut and abrasion-resistant gloves are designed for high-performance protection in demanding environments. Constructed from durable Kevlar, they offer reliable hand safety while maintaining comfort and flexibility. The uncoated design provides enhanced tactile sensitivity for various tasks.
  • Cut and abrasion resistant
  • Made from Kevlar primary material
  • Uncoated for enhanced dexterity
  • ANSI/ISEA Cut Resistance Level A3
  • ANSI/ISEA Abrasion Resistance Level 3
  • EN 388 rating 144XD
  • Launderable and reversible design
  • Knit cuff style
Catalog No. 50-007-94581 Supplier MSC Supplier No. 63618219
These cut and abrasion-resistant gloves are designed for high-performance protection in demanding environments. Constructed from durable Kevlar, they offer reliable hand safety while maintaining comfort and flexibility. The uncoated design provides enhanced tactile sensitivity for various tasks.
  • Cut and abrasion resistant
  • Made from Kevlar primary material
  • Uncoated for enhanced dexterity
  • ANSI/ISEA Cut Resistance Level A3
  • ANSI/ISEA Abrasion Resistance Level 3
  • EN 388 rating 144XD
  • Launderable and reversible design
  • Knit cuff style