Learn More
MSC Ansell Cut-Resistant Gloves, ANSI A4, Nitrile/HPPE, 3X-Large, 1 Pair
Catalog No. 5000803730
Encompass
Ansell Edge cut-resistant gloves provide superior protection for various applications. They feature a durable construction with a nitrile palm coating and a blend of HPPE, polyester, and spandex lining, designed for enhanced comfort and grip.
  • Cut-resistant glove type
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA abrasion resistance level 6
  • Nitrile coating material with palm coverage
  • Primary material includes HPPE, polyester, and spandex
  • Overall length of 13-1/2 in
  • Knit style with rough grip surface
  • Gauntlet cuff style
  • Certified to EN-407 standard
Catalog No. 50-008-03730 Supplier MSC Supplier No. 72859465
Ansell Edge cut-resistant gloves provide superior protection for various applications. They feature a durable construction with a nitrile palm coating and a blend of HPPE, polyester, and spandex lining, designed for enhanced comfort and grip.
  • Cut-resistant glove type
  • ANSI/ISEA cut resistance level A4
  • ANSI/ISEA abrasion resistance level 6
  • Nitrile coating material with palm coverage
  • Primary material includes HPPE, polyester, and spandex
  • Overall length of 13-1/2 in
  • Knit style with rough grip surface
  • Gauntlet cuff style
  • Certified to EN-407 standard