Learn More
MSC Ansell Disposable Gloves, Latex, Medical, 11 mil, Medium, 50 Pack
Catalog No. 5000794139
Encompass
These disposable gloves are designed for medical applications, offering an ambidextrous fit and a grain grip on the fingers for enhanced dexterity. They are non-sterile and blue in color, measuring 12.0 in. in length.
  • Three times greater thickness for snag- and tear-resistant latex exam gloves.
  • Soft latex formulation provides a more elastic fit and superior comfort.
  • Certified performance; NFPA 1999 certified and chemotherapy drug-tested.
Catalog No. 50-007-94139 Supplier MSC Supplier No. 48446884
These disposable gloves are designed for medical applications, offering an ambidextrous fit and a grain grip on the fingers for enhanced dexterity. They are non-sterile and blue in color, measuring 12.0 in. in length.
  • Three times greater thickness for snag- and tear-resistant latex exam gloves.
  • Soft latex formulation provides a more elastic fit and superior comfort.
  • Certified performance; NFPA 1999 certified and chemotherapy drug-tested.