Learn More
MSC Ansell Disposable Gloves, Latex, Powder-Free, Medical, Large, 9.8 mil, 50/Pk.
Catalog No. 5000673478
Encompass
Ansell Microflex Ultra One disposable gloves are medical-grade, powder-free latex gloves designed for durability and comfort. With a thickness of 9.8 mil and a length of 11.8 in., these ambidextrous gloves feature a grain grip on the fingers for enhanced handling.
  • Extended cuff offers additional forearm and wrist protection.
  • Thick finger and palm design provides strong, durable medical exam gloves.
  • Soft latex composition ensures an elastic fit and comfortable feel.
Catalog No. 50-006-73478 Supplier MSC Supplier No. 74176157
Ansell Microflex Ultra One disposable gloves are medical-grade, powder-free latex gloves designed for durability and comfort. With a thickness of 9.8 mil and a length of 11.8 in., these ambidextrous gloves feature a grain grip on the fingers for enhanced handling.
  • Extended cuff offers additional forearm and wrist protection.
  • Thick finger and palm design provides strong, durable medical exam gloves.
  • Soft latex composition ensures an elastic fit and comfortable feel.