Learn More
MSC Ansell Disposable Gloves, Microflex Nitrile, Small, Powder-Free, 100/Pk.
Catalog No. 5000795105
Encompass
These disposable gloves from the Microflex series are designed for general purpose use. They are made of nitrile, are powder-free, and come in a cobalt blue color, making them suitable for various tasks requiring hand protection.
  • Unique cobalt blue color provides contrast.
  • Fully textured for enhanced grip.
  • Versatile for food prep, light-duty cleaning, maintenance, and more.
  • Ambidextrous for easy use.
  • FDA approved and non-sterile.
  • Rolled cuff style.
  • ISO 9001 rated.
Catalog No. 50-007-95105 Supplier MSC Supplier No. 37062205
These disposable gloves from the Microflex series are designed for general purpose use. They are made of nitrile, are powder-free, and come in a cobalt blue color, making them suitable for various tasks requiring hand protection.
  • Unique cobalt blue color provides contrast.
  • Fully textured for enhanced grip.
  • Versatile for food prep, light-duty cleaning, maintenance, and more.
  • Ambidextrous for easy use.
  • FDA approved and non-sterile.
  • Rolled cuff style.
  • ISO 9001 rated.