Learn More
MSC Ansell Disposable Gloves, Microflex, X-Small, Nitrile, 2.8 mil, Powder-Free, 100/Pk.
Catalog No. 5000685982
Encompass
These Ansell Microflex disposable gloves are white, 9.6 in. long, and feature a grain (fingers) grip for enhanced dexterity. They are ambidextrous, FDA approved, and non-sterile.
  • Hydrasoft™ skin moisturizing technology helps prevent dryness.
  • Soft nitrile formulation provides a comfortable fit and feel.
  • Thin and lightweight to ensure excellent tactility.
Catalog No. 50-006-85982 Supplier MSC Supplier No. 69859197
These Ansell Microflex disposable gloves are white, 9.6 in. long, and feature a grain (fingers) grip for enhanced dexterity. They are ambidextrous, FDA approved, and non-sterile.
  • Hydrasoft™ skin moisturizing technology helps prevent dryness.
  • Soft nitrile formulation provides a comfortable fit and feel.
  • Thin and lightweight to ensure excellent tactility.