Ansell TouchNTuff 93-800 disposable gloves are engineered for advanced chemical protection, featuring an innovative multi-layer design with high acetone resistance. These gloves offer enhanced mechanical properties and a bio-based formulation, making them a reliable and sustainable choice for various high-risk applications.
- Reliable protection: ≥15 minutes acetone resistance and superior ketone protection.
- Innovative multi-layer design: MICROCHEM™ Chemical Barrier Technology with Type A chemical resistance.
- Enhanced mechanical properties: EN388 2110A certified for abrasion (level 2) and cut (ISO Cut A) protection.
- High visibility orange color aids in foreign object detection.
- Flexible formulation provides improved fit, feel, and flexibility.
- TÜV certified bio-based formulation contains over 60% bio-based carbon for sustainability.