Learn More
MSC Ansell Disposable Gloves, Nitrile, 8 mil, Small, 50/Pack
Catalog No. 5000802086
Encompass
These disposable nitrile gloves provide an ultra-secure grip due to their Mega Texture surface, making them suitable for handling wet, oily, or fatty objects. With a robust 0.2 mm (8 mil) thickness, they offer tear resistance and are certified for food handling. Their generous fit allows for comfortable double donning, and an extended cuff provides additional forearm and wrist protection.
  • Ultra-secure grip provided by Mega Texture surface
  • Robust 0.2 mm (8 mil) thickness resists tears and protects products
  • Compliant for a wide range of food handling applications
  • Extended cuff design offers added forearm and wrist protection
  • Generous fit allows comfortable double donning over other gloves
Catalog No. 50-008-02086 Supplier MSC Supplier No. 84851518
These disposable nitrile gloves provide an ultra-secure grip due to their Mega Texture surface, making them suitable for handling wet, oily, or fatty objects. With a robust 0.2 mm (8 mil) thickness, they offer tear resistance and are certified for food handling. Their generous fit allows for comfortable double donning, and an extended cuff provides additional forearm and wrist protection.
  • Ultra-secure grip provided by Mega Texture surface
  • Robust 0.2 mm (8 mil) thickness resists tears and protects products
  • Compliant for a wide range of food handling applications
  • Extended cuff design offers added forearm and wrist protection
  • Generous fit allows comfortable double donning over other gloves