Learn More
MSC Ansell Disposable Gloves, Nitrile, Medical, Large, 4.5 mil, 50/Pk.
Catalog No. 5000789490
Encompass
The Ansell Microflex FreeForm series disposable gloves are made from Nitrile, with a nominal palm thickness of 4.5 mil. These medical-grade gloves are designed for high-speed applications, featuring an extended cuff and textured fingertips for enhanced grip, making them suitable for laboratory and medical environments.
  • Textured fingertips for enhanced grip
  • Non-stick properties resist tape and adhesives
  • Added protection with an extended cuff and NFPA 1999 certification
  • Medical grade, powder-free, non-sterile
  • Ambidextrous design
  • Complies with multiple standards including FDA, ASTM, and NFPA
Catalog No. 50-007-89490 Supplier MSC Supplier No. 74169756
The Ansell Microflex FreeForm series disposable gloves are made from Nitrile, with a nominal palm thickness of 4.5 mil. These medical-grade gloves are designed for high-speed applications, featuring an extended cuff and textured fingertips for enhanced grip, making them suitable for laboratory and medical environments.
  • Textured fingertips for enhanced grip
  • Non-stick properties resist tape and adhesives
  • Added protection with an extended cuff and NFPA 1999 certification
  • Medical grade, powder-free, non-sterile
  • Ambidextrous design
  • Complies with multiple standards including FDA, ASTM, and NFPA