Learn More
MSC Ansell Disposable Gloves, Nitrile, Medium, Powder-Free, General Purpose, 100/Pk.
Catalog No. 5000801982
Encompass
These Ansell Microflex disposable gloves are constructed from nitrile, offering a powder-free and ambidextrous design for general purpose use. Featuring a cobalt blue color, a 9.5 in length, and a fully textured grip, these gloves are FDA approved and non-sterile, suitable for various professional applications.
  • Nitrile construction
  • Powder-free design
  • Medium size
  • Thickness of 3.9 mil
  • General purpose grade
  • Rolled cuff style
  • Fully textured surface for enhanced grip
  • Unique cobalt blue color provides contrast
  • Ambidextrous design
  • FDA approved and non-sterile
  • Versatile for food preparation, light-duty cleaning, and maintenance
Catalog No. 50-008-01982 Supplier MSC Supplier No. 37062247
These Ansell Microflex disposable gloves are constructed from nitrile, offering a powder-free and ambidextrous design for general purpose use. Featuring a cobalt blue color, a 9.5 in length, and a fully textured grip, these gloves are FDA approved and non-sterile, suitable for various professional applications.
  • Nitrile construction
  • Powder-free design
  • Medium size
  • Thickness of 3.9 mil
  • General purpose grade
  • Rolled cuff style
  • Fully textured surface for enhanced grip
  • Unique cobalt blue color provides contrast
  • Ambidextrous design
  • FDA approved and non-sterile
  • Versatile for food preparation, light-duty cleaning, and maintenance