Learn More
MSC Ansell Disposable Gloves, Vinyl, Large, Powder-Free, 2.8 mil, 100/Pk.
Catalog No. 5000677318
Encompass
These disposable gloves are from the Microflex series, designed for food handling and general maintenance. They offer vinyl hand protection at a cost-effective rate. The gloves are blue, 9.5 in. long, and feature a smooth grip for a more natural fit. They are ambidextrous and non-sterile.
  • Smooth finish for a more natural fit
  • Vinyl hand protection at a cost-effective rate
  • Excellent for food prep and general maintenance
  • Primary material: vinyl
  • Thickness: 2.8 mil
  • Glove size: large
  • Powder-free
  • Non-sterile
  • Cuff style: rolled
  • Ambidextrous
  • Length: 9.5 in.
  • Standards: EN 420:2003+A1:2009; FDA 21 CFR 177.2600
  • ISO Rating: ISO 9001
  • Country of origin: China
Catalog No. 50-006-77318 Supplier MSC Supplier No. 38620829
These disposable gloves are from the Microflex series, designed for food handling and general maintenance. They offer vinyl hand protection at a cost-effective rate. The gloves are blue, 9.5 in. long, and feature a smooth grip for a more natural fit. They are ambidextrous and non-sterile.
  • Smooth finish for a more natural fit
  • Vinyl hand protection at a cost-effective rate
  • Excellent for food prep and general maintenance
  • Primary material: vinyl
  • Thickness: 2.8 mil
  • Glove size: large
  • Powder-free
  • Non-sterile
  • Cuff style: rolled
  • Ambidextrous
  • Length: 9.5 in.
  • Standards: EN 420:2003+A1:2009; FDA 21 CFR 177.2600
  • ISO Rating: ISO 9001
  • Country of origin: China