Learn More
MSC Ansell Disposable Gloves, X-Small, 2.8 mil, Nitrile, Powder-Free, 250/Pack
Catalog No. 5000765839
Encompass
These nitrile disposable gloves are designed for general-purpose applications, offering an X-Small size with a 2.8 mil thickness. They are powder-free, ambidextrous, and provide a grain (fingers) grip.
  • Primary material: Nitrile
  • Thickness: 2.8 mil
  • Grip surface: Grain (fingers)
  • General purpose grade
  • Non-sterile and powder-free
  • Length: 9.5 in.
  • Ambidextrous
  • FDA approved
  • Static dissipative
  • Blue color
Catalog No. 50-007-65839 Supplier MSC Supplier No. 94461852
These nitrile disposable gloves are designed for general-purpose applications, offering an X-Small size with a 2.8 mil thickness. They are powder-free, ambidextrous, and provide a grain (fingers) grip.
  • Primary material: Nitrile
  • Thickness: 2.8 mil
  • Grip surface: Grain (fingers)
  • General purpose grade
  • Non-sterile and powder-free
  • Length: 9.5 in.
  • Ambidextrous
  • FDA approved
  • Static dissipative
  • Blue color