Learn More
MSC Ansell Exam Gloves, Nitrile, X-Small, 4.3 mil, Powder-Free, 100/Pk.
Catalog No. 5000686261
Encompass
MICROFLEX™ Supreno™ SE SU-690 exam gloves are made with a strong nitrile formulation, offering robust personal protection. Textured fingertips ensure better grip when handling tools and equipment. A low 0.65 AQL provides excellent barrier protection against potentially hazardous substances.
  • Robust nitrile formulation for durable personal protection
  • Advanced barrier protection with low 0.65 AQL for allowable pinholes
  • Textured fingertips allow for assured grip in diverse conditions
Catalog No. 50-006-86261 Supplier MSC Supplier No. 69859221
MICROFLEX™ Supreno™ SE SU-690 exam gloves are made with a strong nitrile formulation, offering robust personal protection. Textured fingertips ensure better grip when handling tools and equipment. A low 0.65 AQL provides excellent barrier protection against potentially hazardous substances.
  • Robust nitrile formulation for durable personal protection
  • Advanced barrier protection with low 0.65 AQL for allowable pinholes
  • Textured fingertips allow for assured grip in diverse conditions