Learn More
MSC Ansell Gloves, Nitrile, Small, 3.5 mil, Powder-Free, Medical Grade, 100/Pk.
Catalog No. 5000794078
Encompass
The Ansell N641 Microflex Onyx disposable gloves are designed for comfort and protection in medical and laboratory settings. These powder-free, medical-grade nitrile gloves are ambidextrous and provide a reliable barrier for various tasks.
  • Chemical splash protection
  • Soft nitrile formulation for comfort and durability
  • Deep black color for contrast and stain hiding
  • Non-sterile for general purpose use
  • Ambidextrous design for easy donning
Catalog No. 50-007-94078 Supplier MSC Supplier No. 39100326
The Ansell N641 Microflex Onyx disposable gloves are designed for comfort and protection in medical and laboratory settings. These powder-free, medical-grade nitrile gloves are ambidextrous and provide a reliable barrier for various tasks.
  • Chemical splash protection
  • Soft nitrile formulation for comfort and durability
  • Deep black color for contrast and stain hiding
  • Non-sterile for general purpose use
  • Ambidextrous design for easy donning