Learn More
Enterprise Technology Solutions Ansell Healthcare Products Coveralls,A40,ZipperFront,f/Lqud/ParticleProtection,3XL,25/CT,WE
SDP
Catalog No. 5000828157
Encompass_Preferred
A40 Liquid and Particle Protection Coveralls with a zippered front protect against hazardous particulates, hazardous aerosols and low-hazard liquid splash. They pass ASTM F1670 and 1671 for blood penetration and bloodborne pathogens. This protective wear also passes ASTM F903 for isocyanate paint and potassium chromate. Microporous-film laminate fabric offers breathability and comfort. Human-centric Reflex design allows better fit and freedom of movement during difficult jobs and projects. When used in nonhazardous applications, size 3XL coveralls are eligible for recycling through The Right Cycle Program.
Catalog No. 50-008-28157 Supplier Enterprise Technology Solutions Supplier No. ANS44306
A40 Liquid and Particle Protection Coveralls with a zippered front protect against hazardous particulates, hazardous aerosols and low-hazard liquid splash. They pass ASTM F1670 and 1671 for blood penetration and bloodborne pathogens. This protective wear also passes ASTM F903 for isocyanate paint and potassium chromate. Microporous-film laminate fabric offers breathability and comfort. Human-centric Reflex design allows better fit and freedom of movement during difficult jobs and projects. When used in nonhazardous applications, size 3XL coveralls are eligible for recycling through The Right Cycle Program.