Learn More
Enterprise Technology Solutions Ansell Healthcare Products Coveralls,Zipper,Elastic Hood/Back/Wrist/Ankle,2X,25/CT,WE
SDP
Catalog No. 5000828120
Encompass_Preferred
A30 Breathable Splash and Particle Protection Coveralls protect against hazardous dry particles, aerosols and low-hazardous limited liquid splash. Air-permeable, SMS fabric provides breathability and comfort and is compliant with NFPA99 for anti-static materials. Design features a zippered front with a 1" flap and elastic on back, wrists, ankles and hood. Human-centric Reflex design allows better fit and freedom of movement. When used in nonhazardous applications, they are eligible for recycling through The Right Cycle Program.
Catalog No. 50-008-28120 Supplier Enterprise Technology Solutions Supplier No. ANS46115
A30 Breathable Splash and Particle Protection Coveralls protect against hazardous dry particles, aerosols and low-hazardous limited liquid splash. Air-permeable, SMS fabric provides breathability and comfort and is compliant with NFPA99 for anti-static materials. Design features a zippered front with a 1" flap and elastic on back, wrists, ankles and hood. Human-centric Reflex design allows better fit and freedom of movement. When used in nonhazardous applications, they are eligible for recycling through The Right Cycle Program.