Learn More
Enterprise Technology Solutions Ansell Healthcare Products Coveralls,Zipper,Elastic Hood/Back/Wrist/Ankle,XL,24/CT,WE
SDP
Catalog No. 5000828159
Encompass_Preferred
A20 Breathable Particle Protection Coveralls protect against hazardous dry particles and feature a zippered front plus elastic hood, back, wrists and ankles. Air-permeable SMMMS fabric provides five layers of stacked barrier protection. Seamless front offers extra protection to the primary exposure area. Popular choices for use include dusty environments such as laboratories and remediation as well as pesticide applications and decontamination modesty garments. Human-centric Reflex design allows better fit and freedom of movement. When used in nonhazardous applications, size XLarge coveralls are eligible for recycling through The Right Cycle Program.
Catalog No. 50-008-28159 Supplier Enterprise Technology Solutions Supplier No. ANS49114
A20 Breathable Particle Protection Coveralls protect against hazardous dry particles and feature a zippered front plus elastic hood, back, wrists and ankles. Air-permeable SMMMS fabric provides five layers of stacked barrier protection. Seamless front offers extra protection to the primary exposure area. Popular choices for use include dusty environments such as laboratories and remediation as well as pesticide applications and decontamination modesty garments. Human-centric Reflex design allows better fit and freedom of movement. When used in nonhazardous applications, size XLarge coveralls are eligible for recycling through The Right Cycle Program.