Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,Anti-Fog, Black Frame/CL Lens
SDP
Catalog No. 5000828582
Encompass_Preferred
Nemesis Safety Glasses provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate clear lenses with anti-fog coating offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.
Catalog No. 50-008-28582 Supplier Enterprise Technology Solutions Supplier No. ANS25679
Nemesis Safety Glasses provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate clear lenses with anti-fog coating offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.