Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,Anti-Fog,Black Frame/Smoke
SDP
Catalog No. 5000827862
Encompass_Preferred
V30 Maverick Safety Glasses offer a durable, lightweight construction coupled with a sleek, modern look. Polycarbonate, smoke-colored lenses with anti-fog coating provide 99.9 percent UVA/UVB/UVC protection. With integrated side shields and brow guard, safety glasses meet optical and impact-protector requirements of the ANSI Z87.1+ standard. Comfort touch points on the temples and nose bridge effectively combine comfort, function and uncompromised protection. Unisex safety eyewear are Recyclable through The Right Cycle Program.
Catalog No. 50-008-27862 Supplier Enterprise Technology Solutions Supplier No. ANS49311
V30 Maverick Safety Glasses offer a durable, lightweight construction coupled with a sleek, modern look. Polycarbonate, smoke-colored lenses with anti-fog coating provide 99.9 percent UVA/UVB/UVC protection. With integrated side shields and brow guard, safety glasses meet optical and impact-protector requirements of the ANSI Z87.1+ standard. Comfort touch points on the temples and nose bridge effectively combine comfort, function and uncompromised protection. Unisex safety eyewear are Recyclable through The Right Cycle Program.