Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,Anti-Scratch,Smoke
SDP
Catalog No. 5000827874
Encompass_Preferred
Elite Safety Glasses provide unmatched protection during both work and play. They offer 99.9 percent UVA/UVB/UVC protection to help prevent eye damage and vision loss. Retro, lightweight design works for a variety of environments, whether you're doing yard work or driving. Anti-scratch, dual lenses offer wraparound protection and wide temples for structural integrity and stability. Protective, unisex glasses are impact-rated and meet ANSIZ87.1+ standards.
Catalog No. 50-008-27874 Supplier Enterprise Technology Solutions Supplier No. ANS21303
Elite Safety Glasses provide unmatched protection during both work and play. They offer 99.9 percent UVA/UVB/UVC protection to help prevent eye damage and vision loss. Retro, lightweight design works for a variety of environments, whether you're doing yard work or driving. Anti-scratch, dual lenses offer wraparound protection and wide temples for structural integrity and stability. Protective, unisex glasses are impact-rated and meet ANSIZ87.1+ standards.