Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,BK Frame/Blue Mirror Lens
SDP
Catalog No. 5000827850
Encompass_Preferred
Nemesis Safety Glasses with a mirror coating provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate blue lenses offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.
Catalog No. 50-008-27850 Supplier Enterprise Technology Solutions Supplier No. ANS14481
Nemesis Safety Glasses with a mirror coating provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate blue lenses offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.