Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,Flex-Point,BK/Smoke Lens
SDP
Catalog No. 5000828156
Encompass_Preferred
Hellraiser Safety Glasses provide comfort and style without sacrificing eye protection. High-performance polycarbonate lenses give 99.9 percent UV protection (UVA/UVB/UVC) and increased durability. Unisex protection glasses meet ANSIZ87.1+ standards. Single-lens wraparound protection features a scratch-resistant lens. Comfort and stability are a given with flex-point temples. Lightweight glasses are ideal for use in a variety of work environments, including industrial manufacturing.
Catalog No. 50-008-28156 Supplier Enterprise Technology Solutions Supplier No. ANS25714
Hellraiser Safety Glasses provide comfort and style without sacrificing eye protection. High-performance polycarbonate lenses give 99.9 percent UV protection (UVA/UVB/UVC) and increased durability. Unisex protection glasses meet ANSIZ87.1+ standards. Single-lens wraparound protection features a scratch-resistant lens. Comfort and stability are a given with flex-point temples. Lightweight glasses are ideal for use in a variety of work environments, including industrial manufacturing.