Learn More
Enterprise Technology Solutions Ansell Healthcare Products Glasses,Safety,UV Protection,No-Slip,BK Frame/Smoke/Mirror
SDP
Catalog No. 5000827859
Encompass_Preferred
Nemesis Safety Glasses provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate smoke-colored lenses with mirror coating offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.
Catalog No. 50-008-27859 Supplier Enterprise Technology Solutions Supplier No. ANS25688
Nemesis Safety Glasses provide effective eye protection and help prevent future eye damage and vision loss. Polycarbonate smoke-colored lenses with mirror coating offer 99.9 percent UV protection (UVA/UVB/UVC). Wraparound frames feature a base curve of eight and extend the rage of peripheral protection. Flexible nose piece design prevents slippage by channeling sweat away from the eyes. Included neck cord secures glasses to you. Sleek and modern safety glasses meet ANSIZ87.1+ standards and are ideal for construction, manufacturing and many other industries.