Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Nitrile,PF,Textured Tips,4.3mil,9.5"L,Large,200/BX,PE
SDP
Catalog No. 5000828544
Encompass_Preferred
Vista Nitrile Exam Gloves offer ultimate comfort and effective cleanliness. 4.3 mil thick nitrile gloves provide reliable protection. Because they're ambidextrous, you can quickly grab a single glove for either hand. The 9.5" long exam gloves are powder-free and are not made with natural rubber latex . They have an AQL of 1.5, exceeding the ASTM AQL 2.5 standard. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves meet U.S. FDA regulations for food-contact applications and are tested for use with chemotherapy drugs. Exam gloves can be recycled through The Right Cycle Program.
Catalog No. 50-008-28544 Supplier Enterprise Technology Solutions Supplier No. ANS62828
Vista Nitrile Exam Gloves offer ultimate comfort and effective cleanliness. 4.3 mil thick nitrile gloves provide reliable protection. Because they're ambidextrous, you can quickly grab a single glove for either hand. The 9.5" long exam gloves are powder-free and are not made with natural rubber latex . They have an AQL of 1.5, exceeding the ASTM AQL 2.5 standard. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves meet U.S. FDA regulations for food-contact applications and are tested for use with chemotherapy drugs. Exam gloves can be recycled through The Right Cycle Program.