Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Nitrile,PF,Textured Tips,5.9mil,9.5"L,Large,100/BX,PE
SDP
Catalog No. 5000828161
Encompass_Preferred
Purple Nitrile Exam Gloves are made from highly durable nitrile polymer and protect against incidental splashes. 5.9 mil thick nitrile gloves provide reliable protection. Because 9.5" long gloves are ambidextrous, you can quickly grab a single glove for either hand. Gloves are powder-free are not made with natural rubber latex. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves are cleared for use in chemotherapy treatments and have an AQL of 1.0, exceeding the ASTM AWL 2.5 standard. Exam gloves can be recycled through The Right Cycle Program.
Catalog No. 50-008-28161 Supplier Enterprise Technology Solutions Supplier No. ANS55083
Purple Nitrile Exam Gloves are made from highly durable nitrile polymer and protect against incidental splashes. 5.9 mil thick nitrile gloves provide reliable protection. Because 9.5" long gloves are ambidextrous, you can quickly grab a single glove for either hand. Gloves are powder-free are not made with natural rubber latex. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves are cleared for use in chemotherapy treatments and have an AQL of 1.0, exceeding the ASTM AWL 2.5 standard. Exam gloves can be recycled through The Right Cycle Program.