Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Nitrile,PF,Textured Tips,6mil,9.5"L,Med,100/BX,BE
SDP
Catalog No. 5000827878
Encompass_Preferred
G10 2Pro Nitrile Gloves allow maximum productivity and safety in many different industries. 6 mil thick gloves provide reliable protection. Because 9.5" long gloves are ambidextrous, you can quickly grab a single glove for either hand. Gloves are powder-free, touch screen-compatible and are not made with natural rubber latex. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves meet U.S. FDA regulations for food-contact applications and can be recycled through The Right Cycle Program.
Catalog No. 50-008-27878 Supplier Enterprise Technology Solutions Supplier No. ANS54422
G10 2Pro Nitrile Gloves allow maximum productivity and safety in many different industries. 6 mil thick gloves provide reliable protection. Because 9.5" long gloves are ambidextrous, you can quickly grab a single glove for either hand. Gloves are powder-free, touch screen-compatible and are not made with natural rubber latex. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Beaded cuffs make gloves easier to don and prevent roll-down. Gloves meet U.S. FDA regulations for food-contact applications and can be recycled through The Right Cycle Program.