Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Nitrile,Textured Tips,M,6.3mil,12"L,100/Bag,10/CT,WE
SDP
Catalog No. 5000827879
Encompass_Preferred
G3 NxT Nitrile Gloves provide safety, security and high levels of cleanliness in a range of environments. Gloves are rigorously tested to ensure regulatory compliance and suitable for ISO Class Three or higher cleanroom environments. According to EU regulations, the gloves are designated as PPE Cat III. Synthetic nitrile polymer allows a flawless fit and unmatched reliability. Textured glove fingertips improve grip and extreme water tightness results in a lower risk of pinholes. Because gloves are ambidextrous, you can quickly grab a single glove for either hand. Beaded cuff makes donning gloves easier while added strength reduces the chances of material tears. Gloves are latex-free, silicone-free, powder-free and static-tested to protect both the wearer and applicant. Cleanroom-compatible polyethylene packaging is provided.
Catalog No. 50-008-27879 Supplier Enterprise Technology Solutions Supplier No. ANS62992
G3 NxT Nitrile Gloves provide safety, security and high levels of cleanliness in a range of environments. Gloves are rigorously tested to ensure regulatory compliance and suitable for ISO Class Three or higher cleanroom environments. According to EU regulations, the gloves are designated as PPE Cat III. Synthetic nitrile polymer allows a flawless fit and unmatched reliability. Textured glove fingertips improve grip and extreme water tightness results in a lower risk of pinholes. Because gloves are ambidextrous, you can quickly grab a single glove for either hand. Beaded cuff makes donning gloves easier while added strength reduces the chances of material tears. Gloves are latex-free, silicone-free, powder-free and static-tested to protect both the wearer and applicant. Cleanroom-compatible polyethylene packaging is provided.