Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Safety,Foam Nitrile,Nylon,Large,12/PK,BK/BE
SDP
Catalog No. 5000827877
Encompass_Preferred
G40 Multi-Purpose Work Gloves provide improved comfort and productivity using seamless-knit nylon with a foam-nitrile coating. Gloves are ideal for tough jobs where strength, grip and dexterity are a must. Protective wear offers excellent dexterity and features improved oil and industrial grease grip. Gloves also maintain the highest abrasion rating (EN388) through at least five wash cycles. Any job or project becomes easier and safer with G40 Work Gloves.
Catalog No. 50-008-27877 Supplier Enterprise Technology Solutions Supplier No. ANS40227
G40 Multi-Purpose Work Gloves provide improved comfort and productivity using seamless-knit nylon with a foam-nitrile coating. Gloves are ideal for tough jobs where strength, grip and dexterity are a must. Protective wear offers excellent dexterity and features improved oil and industrial grease grip. Gloves also maintain the highest abrasion rating (EN388) through at least five wash cycles. Any job or project becomes easier and safer with G40 Work Gloves.