Learn More
Enterprise Technology Solutions Ansell Healthcare Products Safety Gloves, Nitrile Foam Coating, Medium, 2/PR, GY/WE
SDP
Catalog No. 5000827872
Encompass_Preferred
Knit-lined, dipped, foam gloves provide comfort and protection. Patented foam coating is resistant to snags and abrasions and provides excellent grip in light oil applications by channeling the oil away from the glove surface. Ideal for automotive assembly and small parts handling. Ergonomic, formfitting liner design allows the glove to mold naturally to the hand, minimizing fatigue. Exceptional comfort and fit increase worker acceptance and safety compliance.
Catalog No. 50-008-27872 Supplier Enterprise Technology Solutions Supplier No. ANS118008
Knit-lined, dipped, foam gloves provide comfort and protection. Patented foam coating is resistant to snags and abrasions and provides excellent grip in light oil applications by channeling the oil away from the glove surface. Ideal for automotive assembly and small parts handling. Ergonomic, formfitting liner design allows the glove to mold naturally to the hand, minimizing fatigue. Exceptional comfort and fit increase worker acceptance and safety compliance.