Learn More
Enterprise Technology Solutions Ansell Healthcare Products Safety Goggles, w/Face Shield, Indirect Vent, Anti-Fog, BE
SDP
Catalog No. 5000827873
Encompass_Preferred
V90 Safety Goggles with Face Shield provide eye and full-face protection from liquids and particles with less bulk and hassle. Two-in-one splash goggles detach from the face shield mask and can be used separately. Indirect venting allows free airflow while effectively safeguarding against liquids entering the eyes. Goggles fit over most prescription eye wear and provide a soft, pliable frame that molds to the face for increased comfort and a cylindrical lens for increased visibility. Face shield, like the goggles, conforms to the face-shape for added protection and includes an adjustable strap for a snug fit. Safety goggles and face shield provide clear lenses with anti-fog coating.
Catalog No. 50-008-27873 Supplier Enterprise Technology Solutions Supplier No. ANS18629
V90 Safety Goggles with Face Shield provide eye and full-face protection from liquids and particles with less bulk and hassle. Two-in-one splash goggles detach from the face shield mask and can be used separately. Indirect venting allows free airflow while effectively safeguarding against liquids entering the eyes. Goggles fit over most prescription eye wear and provide a soft, pliable frame that molds to the face for increased comfort and a cylindrical lens for increased visibility. Face shield, like the goggles, conforms to the face-shape for added protection and includes an adjustable strap for a snug fit. Safety goggles and face shield provide clear lenses with anti-fog coating.