Learn More
Enterprise Technology Solutions Ansell Healthcare Products Shoe Covers,Elastic,Universal,7"H,100/PK,3PK/CT,WE
SDP
Catalog No. 5000828480
Encompass_Preferred
Made of breathable, microporous film, A40 Liquid and Particle Protection Shoe Covers keep out debris, nonhazardous liquid chemicals and dry particulates. They pass NFPA 99 for anti-static materials and ASTM F1670 for penetration of blood and bodily fluids. Ultra-sonic bonded seams, elastic opening and seamless sole provide extra strength and ensure durability. Use in liquid handling, pharmaceutical manufacturing, general manufacturing and more. One size fits all so shoe covers can be used by most.
Catalog No. 50-008-28480 Supplier Enterprise Technology Solutions Supplier No. ANS27000
Made of breathable, microporous film, A40 Liquid and Particle Protection Shoe Covers keep out debris, nonhazardous liquid chemicals and dry particulates. They pass NFPA 99 for anti-static materials and ASTM F1670 for penetration of blood and bodily fluids. Ultra-sonic bonded seams, elastic opening and seamless sole provide extra strength and ensure durability. Use in liquid handling, pharmaceutical manufacturing, general manufacturing and more. One size fits all so shoe covers can be used by most.