Learn More
MSC Ansell HyFlex 70-215, Cut A3, Abrasion 3, Medium, 1 Pair
Catalog No. 5000677794
Encompass
Ansell HyFlex® 70-215 cut and abrasion-resistant gloves are designed for demanding applications. These gloves feature a durable Kevlar® construction, offering robust protection against cuts and abrasions.
  • Launderable for extended use
  • Reversible design for versatility
  • Knit cuff style for a secure fit
  • ANSI/ISEA Cut Resistance Level A3 for enhanced safety
  • ANSI/ISEA Abrasion Resistance Level 3 for durability
  • Primary material is Kevlar®
  • Uncoated for tactile sensitivity
  • Yellow color for high visibility
  • Medium size (numeric size 8) with a 7 gauge construction
Catalog No. 50-006-77794 Supplier MSC Supplier No. 05966874
Ansell HyFlex® 70-215 cut and abrasion-resistant gloves are designed for demanding applications. These gloves feature a durable Kevlar® construction, offering robust protection against cuts and abrasions.
  • Launderable for extended use
  • Reversible design for versatility
  • Knit cuff style for a secure fit
  • ANSI/ISEA Cut Resistance Level A3 for enhanced safety
  • ANSI/ISEA Abrasion Resistance Level 3 for durability
  • Primary material is Kevlar®
  • Uncoated for tactile sensitivity
  • Yellow color for high visibility
  • Medium size (numeric size 8) with a 7 gauge construction