Learn More
MSC Ansell HyFlex 70-225, Cut A3/Abrasion 3, Medium, Kevlar, 1 Pair
Catalog No. 5000802922
Encompass
Ansell HyFlex® 70-225 gloves are designed to provide advanced cut and abrasion resistance for various industrial applications. These gloves feature a seamless knit Kevlar construction, offering practical durability and comfort.
  • Offers ANSI/ISEA A3 cut resistance for enhanced safety
  • Provides ANSI/ISEA 3 abrasion resistance for durability
  • Constructed from Kevlar for superior protection
  • Features an uncoated design with a textured grip for improved handling
  • Reversible design for extended glove life
  • Knit cuff ensures a secure and comfortable fit
Catalog No. 50-008-02922 Supplier MSC Supplier No. 87929238
Ansell HyFlex® 70-225 gloves are designed to provide advanced cut and abrasion resistance for various industrial applications. These gloves feature a seamless knit Kevlar construction, offering practical durability and comfort.
  • Offers ANSI/ISEA A3 cut resistance for enhanced safety
  • Provides ANSI/ISEA 3 abrasion resistance for durability
  • Constructed from Kevlar for superior protection
  • Features an uncoated design with a textured grip for improved handling
  • Reversible design for extended glove life
  • Knit cuff ensures a secure and comfortable fit