Learn More
MSC Ansell Kimtech Sterling Nitrile-Xtra Gloves, Medium, 3.5 mil, Powder-Free, 100/Box
Catalog No. 5000794507
Encompass
Kimtech Sterling Nitrile-Xtra Exam Gloves provide exceptional dexterity, comfort, and protection for laboratory applications. Made from nitrile, these powder-free gloves combine the sensitivity of latex with the strength of nitrile.
  • Optimal dexterity, comfort, and protection for laboratory work.
  • Tested against chemotherapy drugs (ASTM D6978) and NFPA 1999 certified.
  • Advanced polymer performance provides latex-like sensitivity with nitrile strength.
  • Enhanced dexterity from textured fingertips and ambidextrous fit.
  • Superior quality with AQL 1.0 for robust barrier and tactile sensitivity.
  • Extended 12 in cuff provides chemical splash protection.
  • Features 3.5 mil thickness; powder-free, non-sterile, gray color.
  • Offers 9.5 in length.
Catalog No. 50-007-94507 Supplier MSC Supplier No. 48381107
Kimtech Sterling Nitrile-Xtra Exam Gloves provide exceptional dexterity, comfort, and protection for laboratory applications. Made from nitrile, these powder-free gloves combine the sensitivity of latex with the strength of nitrile.
  • Optimal dexterity, comfort, and protection for laboratory work.
  • Tested against chemotherapy drugs (ASTM D6978) and NFPA 1999 certified.
  • Advanced polymer performance provides latex-like sensitivity with nitrile strength.
  • Enhanced dexterity from textured fingertips and ambidextrous fit.
  • Superior quality with AQL 1.0 for robust barrier and tactile sensitivity.
  • Extended 12 in cuff provides chemical splash protection.
  • Features 3.5 mil thickness; powder-free, non-sterile, gray color.
  • Offers 9.5 in length.