Learn More
MSC Ansell Latex Medical Examination Gloves, X-Large, Textured, Powder-Free, 1000/Case
Catalog No. 5000688429
Encompass
The MICROFLEX™ Evolution One™ EV-2050 is a latex medical exam glove designed to provide reliable comfort during extended wear. It offers secure protection and features a fully textured surface for a secure grip, also suitable for a variety of food handling applications.
  • Comfortable latex provides reliability during extended wear
  • Fully textured surface for secure grip
  • Powder-free for reduced contamination risk
  • Beaded cuff for easy donning and doffing
  • Achieves 1.5 AQL freedom from holes (inspection level I)
  • Finger thickness of 0.15 mm / 5.9 mil
  • Palm thickness of 0.14 mm / 5.5 mil
  • Glove length of 245 mm / 9.65 in
  • Medical examination grade, certified to ASTM D3578, EN ISO 21420:2020, US QSR/FDA 510(k)
  • Suitable for medical examination and food handling
Catalog No. 50-006-88429 Supplier MSC Supplier No. 87663167
The MICROFLEX™ Evolution One™ EV-2050 is a latex medical exam glove designed to provide reliable comfort during extended wear. It offers secure protection and features a fully textured surface for a secure grip, also suitable for a variety of food handling applications.
  • Comfortable latex provides reliability during extended wear
  • Fully textured surface for secure grip
  • Powder-free for reduced contamination risk
  • Beaded cuff for easy donning and doffing
  • Achieves 1.5 AQL freedom from holes (inspection level I)
  • Finger thickness of 0.15 mm / 5.9 mil
  • Palm thickness of 0.14 mm / 5.5 mil
  • Glove length of 245 mm / 9.65 in
  • Medical examination grade, certified to ASTM D3578, EN ISO 21420:2020, US QSR/FDA 510(k)
  • Suitable for medical examination and food handling