Learn More
MSC Ansell Liner Gloves, Knitted, Cotton Blend, Sweat Absorbing, 1 Pair
Catalog No. 5000802396
Encompass
These knitted liner gloves are engineered for professionals and designed to be worn under rubber insulated gloves. They provide sweat absorption and comfort, helping users stay dry, reduce irritation, and enhance performance during extended periods in demanding environments.
  • Enhanced comfort and breathability
  • Sweat management
  • Reduces irritation
  • Recommended as a liner under an unsupported glove
Catalog No. 50-008-02396 Supplier MSC Supplier No. 45265022
These knitted liner gloves are engineered for professionals and designed to be worn under rubber insulated gloves. They provide sweat absorption and comfort, helping users stay dry, reduce irritation, and enhance performance during extended periods in demanding environments.
  • Enhanced comfort and breathability
  • Sweat management
  • Reduces irritation
  • Recommended as a liner under an unsupported glove