Learn More
MSC Ansell MICROFLEX Aloe Sense N96 Gloves, Nitrile, Teal, Non-Sterile, 100/Disp.
Catalog No. 5000677925
Encompass
These gloves offer comfortable, reliable protection with aloe and chamomile. Designed with a proprietary aloe and chamomile inner coating, they prevent skin dryness. The soft nitrile formulation provides a comfortable fit and feel during extended wear.
  • Proprietary aloe and chamomile coating to protect hand health
  • Soft, nitrile composition for comfort during extended wear
  • Textured fingers provide a more secure grip
Catalog No. 50-006-77925 Supplier MSC Supplier No. 18188656
These gloves offer comfortable, reliable protection with aloe and chamomile. Designed with a proprietary aloe and chamomile inner coating, they prevent skin dryness. The soft nitrile formulation provides a comfortable fit and feel during extended wear.
  • Proprietary aloe and chamomile coating to protect hand health
  • Soft, nitrile composition for comfort during extended wear
  • Textured fingers provide a more secure grip