Learn More
MSC Ansell Microflex FreeForm Gloves, Nitrile, 2X-Large, 4.7 mil, Powder-Free, 50/Pack
Catalog No. 5000803146
Encompass
These disposable nitrile examination gloves are blue, 11.4 in long, and feature a grain (fingers) grip. They are ambidextrous, FDA approved, and non-sterile.
  • Textured fingertips for enhanced grip
  • Non-stick properties resist tape and adhesives
  • Added protection with an extended cuff and NFPA 1999 certified
Catalog No. 50-008-03146 Supplier MSC Supplier No. 51183069
These disposable nitrile examination gloves are blue, 11.4 in long, and feature a grain (fingers) grip. They are ambidextrous, FDA approved, and non-sterile.
  • Textured fingertips for enhanced grip
  • Non-stick properties resist tape and adhesives
  • Added protection with an extended cuff and NFPA 1999 certified