Learn More
MSC Ansell MICROFLEX XCEED Nitrile Gloves, Blue, Powder-Free, X-Small, 100/Pack
Catalog No. 5000676488
Encompass
MICROFLEX™ XCEED™ XC-310 gloves are designed with proprietary ERGOFORM™ Technology to support musculoskeletal health and increase worker productivity. These blue nitrile gloves are manufactured without accelerators to reduce the risk of allergic reactions and offer extra durability with a stronger formulation while maintaining tactile sensitivity.
  • Made without accelerators to protect from allergic reactions
  • ERGOFORM™ Technology supports musculoskeletal health
  • Fentanyl-tested for enhanced user protection
  • Extra durability to protect against rips and tears
  • Enhanced comfort with a softer, lightweight feel
  • Smartly packaged with easy, single glove dispensing
  • Strong barrier integrity with 0.65 AQL
  • Nitrile material, powder-free
  • Textured fingers for improved grip
  • Tested for use with chemotherapy drugs
Catalog No. 50-006-76488 Supplier MSC Supplier No. 92891936
MICROFLEX™ XCEED™ XC-310 gloves are designed with proprietary ERGOFORM™ Technology to support musculoskeletal health and increase worker productivity. These blue nitrile gloves are manufactured without accelerators to reduce the risk of allergic reactions and offer extra durability with a stronger formulation while maintaining tactile sensitivity.
  • Made without accelerators to protect from allergic reactions
  • ERGOFORM™ Technology supports musculoskeletal health
  • Fentanyl-tested for enhanced user protection
  • Extra durability to protect against rips and tears
  • Enhanced comfort with a softer, lightweight feel
  • Smartly packaged with easy, single glove dispensing
  • Strong barrier integrity with 0.65 AQL
  • Nitrile material, powder-free
  • Textured fingers for improved grip
  • Tested for use with chemotherapy drugs