Learn More
MSC Ansell Nitrile Exam Gloves, Powder-Free, Medium, Chemotherapy Tested, 200/Box
Catalog No. 5000684232
Encompass
Kimtech™ Sterling™ Nitrile Exam Gloves are manufactured using an innovative synthetic nitrile polymer, offering exceptional dexterity and comfort. They provide certified protection against various contaminants, suitable for laboratory and production environments.
  • Exceptional dexterity and comfort
  • Tested against chemotherapy drugs (ASTM D6978)
  • AQL 0.65 for freedom from holes
  • Latex-free, silicone-free, and powder-free
  • Textured fingertips for enhanced grip
  • Beaded cuffs for strength and easy donning
  • Ambidextrous, gray color
  • PPE Cat III compliant
Catalog No. 50-006-84232 Supplier MSC Supplier No. 66630245
Kimtech™ Sterling™ Nitrile Exam Gloves are manufactured using an innovative synthetic nitrile polymer, offering exceptional dexterity and comfort. They provide certified protection against various contaminants, suitable for laboratory and production environments.
  • Exceptional dexterity and comfort
  • Tested against chemotherapy drugs (ASTM D6978)
  • AQL 0.65 for freedom from holes
  • Latex-free, silicone-free, and powder-free
  • Textured fingertips for enhanced grip
  • Beaded cuffs for strength and easy donning
  • Ambidextrous, gray color
  • PPE Cat III compliant