Learn More
MSC Ansell Nitrile Gloves, Microflex Daily Defense 10-733, Size L, 100/Box
Catalog No. 5000802836
Encompass
These nitrile gloves offer reliable, multi-purpose protection for everyday activities, featuring extra durability. They are medical exam grade, suitable for safely cleaning, disinfecting, and wound treatment, and guard against unseen hazards during cleaning and sanitization of commonly used spaces.
  • Offers protection against viruses, bacteria, and other microorganisms
  • Medical exam grade, ideal for safely cleaning and wound treatment
  • Textured fingertips provide a secure grip
  • Meets ASTM F1671 requirements for protection against viruses, bacteria, and other micro-organisms
  • Genuine versatility for tasks including virus/bacteria protection, first aid, cleaning, and food preparation
  • Powder-free and silicone-free
  • Ambidextrous design
Catalog No. 50-008-02836 Supplier MSC Supplier No. 67378638
These nitrile gloves offer reliable, multi-purpose protection for everyday activities, featuring extra durability. They are medical exam grade, suitable for safely cleaning, disinfecting, and wound treatment, and guard against unseen hazards during cleaning and sanitization of commonly used spaces.
  • Offers protection against viruses, bacteria, and other microorganisms
  • Medical exam grade, ideal for safely cleaning and wound treatment
  • Textured fingertips provide a secure grip
  • Meets ASTM F1671 requirements for protection against viruses, bacteria, and other micro-organisms
  • Genuine versatility for tasks including virus/bacteria protection, first aid, cleaning, and food preparation
  • Powder-free and silicone-free
  • Ambidextrous design