Learn More
MSC Ansell Particulate Respirator, N95, Exhalation Valve, Molded Cup, 10/Box
Catalog No. 5000795462
Encompass
The Ansell KleenGuard particulate respirator 3300 series provides respiratory protection against airborne particles, meeting NIOSH N95 standards. Designed with a molded cup and exhalation valve, it offers reliable protection for various applications.
  • Molded cup construction
  • N95 filter rating
  • Protection against airborne particles
  • Elastic strap type
  • NIOSH Certified N95 standard
  • Includes exhalation valve
Catalog No. 50-007-95462 Supplier MSC Supplier No. 29630407
The Ansell KleenGuard particulate respirator 3300 series provides respiratory protection against airborne particles, meeting NIOSH N95 standards. Designed with a molded cup and exhalation valve, it offers reliable protection for various applications.
  • Molded cup construction
  • N95 filter rating
  • Protection against airborne particles
  • Elastic strap type
  • NIOSH Certified N95 standard
  • Includes exhalation valve