Learn More
MSC Ansell Work Gloves, 40-105 Series, Nitrile-Coated, Medium, 1 Pair
Catalog No. 5000793553
Encompass
These general purpose work gloves are constructed from cotton with a cotton lining and a full nitrile coating, designed for applications requiring hand protection and static dissipation. They feature a gauntlet cuff, wing thumb, and full-fingered coverage, conforming to various international safety standards.
  • General purpose design for various applications including equipment handling and material delivery
  • Constructed from cotton with a cotton lining
  • Fully coated with nitrile on the palm for solid coat protection
  • Gauntlet cuff style and full-fingered coverage
  • Medium size, corresponding to numeric size 8
  • Gray, seamless, and static dissipative
  • Conforms to REACH, CE, EN ISO 21420:2020, CAT II, and EN 388:2016 4121X standards
  • Maximum temperature resistance of 104°F (40°C)
Catalog No. 50-007-93553 Supplier MSC Supplier No. 89421754
These general purpose work gloves are constructed from cotton with a cotton lining and a full nitrile coating, designed for applications requiring hand protection and static dissipation. They feature a gauntlet cuff, wing thumb, and full-fingered coverage, conforming to various international safety standards.
  • General purpose design for various applications including equipment handling and material delivery
  • Constructed from cotton with a cotton lining
  • Fully coated with nitrile on the palm for solid coat protection
  • Gauntlet cuff style and full-fingered coverage
  • Medium size, corresponding to numeric size 8
  • Gray, seamless, and static dissipative
  • Conforms to REACH, CE, EN ISO 21420:2020, CAT II, and EN 388:2016 4121X standards
  • Maximum temperature resistance of 104°F (40°C)