Learn More
MSC Ansell Work Gloves, Edge, X-Large, Nitrile-Coated, HPPE Lined, 1 Pair
Catalog No. 5000788964
Encompass
Ansell Edge Series work gloves are designed for industrial applications requiring superior protection and dexterity. These gloves are ideal for handling sharp objects, machining, and assembly tasks, providing robust performance in demanding environments.
  • X-large size (numeric size 10)
  • Nitrile-coated palm provides rough grip
  • HPPE, spandex, and nylon lining for comfort and protection
  • Knit cuff style for a secure fit
  • High-visibility yellow and black color
  • ANSI/ISEA abrasion resistance level 4
  • Overall length of 10-1/2 in
  • Suitable for general purpose, industrial, mechanic's, and lifting applications
Catalog No. 50-007-88964 Supplier MSC Supplier No. 72859903
Ansell Edge Series work gloves are designed for industrial applications requiring superior protection and dexterity. These gloves are ideal for handling sharp objects, machining, and assembly tasks, providing robust performance in demanding environments.
  • X-large size (numeric size 10)
  • Nitrile-coated palm provides rough grip
  • HPPE, spandex, and nylon lining for comfort and protection
  • Knit cuff style for a secure fit
  • High-visibility yellow and black color
  • ANSI/ISEA abrasion resistance level 4
  • Overall length of 10-1/2 in
  • Suitable for general purpose, industrial, mechanic's, and lifting applications